Learn More
GCNT1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GCNT1.
Supplier:  Abnova Corporation PAB4301
Catalog No. 89-131-352
Add to Cart
Description
Description
This gene is a member of the beta-1,6-N-acetylglucosaminyltransferase gene family. It is essential to the formation of Gal beta 1-3(GlcNAc beta 1-6)GalNAc structures and the core 2 O-glycan branch. The gene coding this enzyme was originally mapped to 9q21, but was later localized to 9q13. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq
Specifications
Specifications
GCNT1
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of GCNT1.
In PBS (0.09% sodium azide)
G6NT_HUMAN
GCNT1
A synthetic peptide (conjugated with KLH) corresponding to internal region of human GCNT1.
400 μL
Primary
Human, Mouse
Liquid
ELISA, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user.
GCNT1
C2GNT/C2GNT-L/C2GNT1/G6NT/MGC126335/MGC126336/NACGT2/NAGCT2
Rabbit
Protein G purification
RUO
2650
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Recommendations
Recommendations