Learn More
GJA1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GJA1.
Manufacturer:  Abnova Corporation PAB12790
Catalog No. 89-137-549
Add to cart
Description
Description
Specifications
Specifications
GJA1
1 mg/mL
Rabbit polyclonal antibody raised against synthetic peptide of GJA1.
In HEPES, 150mM NaCl (50% glycerol, 0.01% BSA, 0.03% sodium azide)
Rabbit
Protein A purification
RUO
Primary
Human, Murine, Rat
Western Blot
Unconjugated
Western Blot (1:1000-1:5000) The optimal working dilution should be determined by the end user.
Liquid
A synthetic peptide corresponding to human GJA1.
100 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal