Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLDN, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of GLDN.

Manufacturer:  Abnova Corporation PAB12523

Catalog No. 89-115-898


Add to cart

Description

Description

Specifications

Specifications

GLDN
Unconjugated
The optimal working dilution should be determined by the end user.
GLDN
CLOM|COLM|CRG-L2|CRGL2|FLJ23917|UNC-112
Rabbit
100 ug
Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Reacts with residues 160-176 [LVGKADEKASEHHSPQA] of human CRG-L2. This sequence is between the collagen domains (residues 29-88 and 89-149) and the olfactomedin domain (residues 189-433).
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of GLDN.
In PBS (0.02% sodium azide)
Liquid
GLDN
A synthetic peptide corresponding to amino acids 160-176 of human GLDN.
RUO
Primary
342035
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only