Learn More
GLP1R Rabbit anti-Human, Monkey, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GLP1R.
Manufacturer:  Abnova Corporation PAB26372
Catalog No. 89-128-012
Add to cart
Description
Description
Specifications
Specifications
GLP1R
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2.5 ug/mL) The optimal working dilution should be determined by the end user.
GLP1R
Liquid
GLP1R
A synthetic peptide corresponding to 16 amino acids at N-terminal extracellular domain of human GLP1R.
50 ug
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against synthetic peptide of GLP1R.
In PBS (0.09% sodium azide)
P43220
MGC138331
Rabbit
Immunoaffinity chromatography
RUO
Primary
2740
Human, Monkey