Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLRX, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of GLRX.

Manufacturer:  Abnova Corporation PAB12423

Catalog No. 89-115-797


Add to cart

Description

Description

Specifications

Specifications

GLRX
Unconjugated
Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
GLRX
Liquid
GLRX
A synthetic peptide corresponding to amino acids 40-100 of human GLRX.
RUO
Primary
2745
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of GLRX.
In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide)
P35754
GRX|GRX1|MGC117407
Rabbit
100 uL
Store at 4°C. Do not freeze.
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only