Learn More
Rabbit anti-Glucose Oxidase, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP244232
Catalog No. NBP244232
Add to cart
Description
Description
Specifically detects Glucose Oxidase in C. elegans, Fungi samples, and it is validated for ELISA, Immunohistochemistry, Western Blotting
Glucose Oxidase Polyclonal antibody specifically detects Glucose Oxidase in C. elegans, Fungi samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
Specifications
Glucose Oxidase
LYOPH
Western Blot 1:3500 - 1:16000, ELISA 1:35000 - 1:160000, Immunohistochemistry 1:2000 - 1:8000
Purified
Rabbit
IgG
0.1 mg
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
C. elegans, Fungi
Western Blot, ELISA, Immunohistochemistry
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Beta D Glucose Oxygen 1 Oxido Reductase, Glucose Oxyhydrase, GOD
Anti-Glucose Oxidase Antibody was produced by repeated immunizations with Aspergillus niger Glucose Oxidase protein.
IgG purified
RUO
Primary
Reconstitute with 100 ul deionized water.