Learn More
GNAZ, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GNAZ.
Manufacturer: Abnova Corporation PAB18387
Description
The protein encoded by this gene is a member of a G protein subfamily that mediates signal transduction in pertussis toxin-insensitive systms. This encoded protein may play a role in maintaining the ionic balance of perilymphatic and endolymphatic cochlear fluids. [provided by RefSeq
Specifications
GNAZ | |
Polyclonal | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:5000) The optimal working dilution should be determined by the end user. | |
GNAZ | |
GNAZ | |
A synthetic peptide corresponding to residues surrounding S16 of human GNAZ. | |
100 μg | |
Primary | |
This antibody is specific to GNAZ. | |
Liquid |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of GNAZ. | |
In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
P19086 | |
Rabbit | |
Affinity purification | |
RUO | |
2781 | |
Human, Mouse, Rat |
Safety and Handling
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only