Learn More
GPBAR1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPBAR1.
Supplier:  Abnova Corporation PAB27696
Catalog No. 89-129-212
Add to Cart
Description
Description
This gene encodes a member of the G protein-coupled receptor (GPCR) superfamily. This enzyme functions as a cell surface receptor for bile acids. Treatment of cells expressing this GPCR with bile acids induces the production of intracellular cAMP, activation of a MAP kinase signaling pathway, and internalization of the receptor. The receptor is implicated in the suppression of macrophage functions and regulation of energy homeostasis by bile acids. Alternative splicing results in multiple transcript variants encoding the same protein. [provided by RefSeq
Specifications
Specifications
GPBAR1
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of GPBAR1.
In PBS (0.09% sodium azide)
Q8TDU6
GPBAR1
A synthetic peptide corresponding to 16 amino acid at C-terminus of human GPBAR1.
50 μg
Primary
BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (11-27 ug/mL) The optimal working dilution should be determined by the end user.
GPBAR1
BG37/GPCR/GPCR19/GPR131/M-BAR/MGC40597/TGR5
Rabbit
Immunoaffinity chromatography
RUO
151306
Human
Liquid
Product Suggestions
Product Suggestions