Learn More
GPR108 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPR108.
Supplier:  Abnova Corporation PAB26367
Catalog No. 89-128-007
Add to Cart
Description
Description
Specifications
Specifications
GPR108
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of GPR108.
In PBS (0.09% sodium azide)
Q9NPR9
GPR108
A synthetic peptide corresponding to 18 amino acids at 1st cytoplasmic domain of human GPR108.
50 μg
Primary
BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) The optimal working dilution should be determined by the end user.
GPR108
LUSTR2/MGC14393
Rabbit
Immunoaffinity chromatography
RUO
56927
Human
Liquid
Recommendations
Recommendations