Learn More
GPR162 Rabbit anti-Dog, Horse, Human, Monkey, Pig, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPR162.
Manufacturer:  Abnova Corporation PAB26292
Catalog No. 89-127-933
Add to cart
Description
Description
This gene was identified upon genomic analysis of a gene-dense region at human chromosome 12p13. It appears to be mainly expressed in the brain; however, its function is not known. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq
Specifications
Specifications
GPR162
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1 ug/mL) The optimal working dilution should be determined by the end user.
GPR162
Liquid
GPR162
A synthetic peptide corresponding to 17 amino acids at 1st extracellular domain of human GPR162.
50 ug
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against synthetic peptide of GPR162.
In PBS (0.09% sodium azide)
Q16538
A-2|GRCA
Rabbit
Immunoaffinity chromatography
RUO
Primary
27239
Canine, Equine, Human, Monkey, Porcine