Learn More
GPR20 Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPR20.
Manufacturer:  Abnova Corporation PAB16197
Catalog No. 89-118-087
Add to cart
Description
Description
Specifications
Specifications
GPR20
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) The optimal working dilution should be determined by the end user.
GPR20
Liquid
Rabbit
Immunoaffinity purification
RUO
Primary
2843
Bovine, Human, Murine, Rat
Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against synthetic peptide of GPR20.
In PBS (0.09% sodium azide)
Q99678
GPR20
A synthetic peptide (conjugated with KLH) corresponding to human GPR20.
50 ug
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
3rd cytoplasmic domain of human. Predicted crossreactivity with mouse and rat due to sequence similarity.