Learn More
GPR27 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPR27.
Manufacturer:  Abnova Corporation PAB26019
Catalog No. 89-127-656
Add to cart
Description
Description
Specifications
Specifications
GPR27
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4 ug/mL) The optimal working dilution should be determined by the end user.
GPR27
Liquid
GPR27
A synthetic peptide corresponding to 19 amino acids at N-terminus of human GPR27.
50 ug
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Immunocytochemistry, Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against synthetic peptide of GPR27.
In PBS (0.09% sodium azide)
Q9NS67
SREB1
Rabbit
Immunoaffinity chromatography
RUO
Primary
2850
Human