Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR4 Rabbit anti-Dog, Human, Monkey, Pig, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic peptide of GPR4.
Manufacturer: Abnova Corporation PAB25964
Specifications
GPR4 | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (15 ug/mL) The optimal working dilution should be determined by the end user. | |
GPR4 | |
Liquid | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
Primary | |
2828 | |
Canine, Human, Monkey, Porcine |
Immunohistochemistry (PFA fixed) | |
Rabbit polyclonal antibody raised against synthetic peptide of GPR4. | |
In PBS (0.09% sodium azide) | |
P46093 | |
GPR4 | |
A synthetic peptide corresponding to 20 amino acids at cytoplasmic domain of human GPR4. | |
50 ug | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only