Learn More
GPR82 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPR82.
Manufacturer:  Abnova Corporation PAB25989
Catalog No. 89-127-624
Add to cart
Description
Description
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins.[supplied by OMIM
Specifications
Specifications
GPR82
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (6-16 ug/mL) The optimal working dilution should be determined by the end user.
GPR82
Liquid
Rabbit
Immunoaffinity chromatography
RUO
Primary
27197
Human
Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against synthetic peptide of GPR82.
In PBS (0.09% sodium azide)
Q96P67
GPR82
A synthetic peptide corresponding to 15 amino acids at cytoplasmic domain of human GPR82.
50 ug
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
BLAST analysis of the peptide immunogen showed no homology with other human proteins.