Learn More
GPRC5A Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPRC5A.
Supplier: Abnova Corporation PAB4823
Description
This gene encodes a member of the type 3 G protein-coupling receptor family, characterized by the signature 7-transmembrane domain motif. The encoded protein may be involved in interaction between retinoid acid and G protein signalling pathways. Retinoic acid plays a critical role in development, cellular growth, and differentiation. This gene may play a role in embryonic development and epithelial cell differentiation. [provided by RefSeq
Specifications
GPRC5A | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of GPRC5A. | |
In PBS (0.09% sodium azide) | |
NP_003970 | |
GPRC5A | |
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human GPRC5A. | |
400 μL | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:1000) Western Blot (1:100-500) The optimal working dilution should be determined by the end user. | |
GPRC5A | |
GPCR5A/RAI3/RAIG1 | |
Rabbit | |
Protein G purification | |
RUO | |
9052 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only