Learn More
GPRC6A Rabbit anti-Human, Monkey, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPRC6A.
Manufacturer:  Abnova Corporation PAB26473
Catalog No. 89-128-112
Add to cart
Description
Description
Specifications
Specifications
GPRC6A
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user.
GPRC6A
Liquid
GPRC6A
A synthetic peptide corresponding to 17 amino acids at N-terminal extracellular domain of human GPRC6A.
50 ug
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against synthetic peptide of GPRC6A.
In PBS (0.09% sodium azide)
Q5T6X5
GPCR|bA86F4.3
Rabbit
Immunoaffinity chromatography
RUO
Primary
222545
Human, Monkey