Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPRIN3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPRIN3.
Manufacturer: Abnova Corporation PAB17725
Specifications
GPRIN3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of GPRIN3. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
Q6ZVF9 | |
GPRIN3 | |
A synthetic peptide corresponding to internal of human GPRIN3. | |
RUO | |
285513 | |
Human |
ELISA, Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:50-1:100) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
GPRIN3 | |
FLJ42625/GRIN3 | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total GPRIN3 protein. | |
Liquid |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only