Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-GRASP65, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236527

Catalog No. NBP236527


Add to cart

Description

Description

GRASP65 Polyclonal antibody specifically detects GRASP65 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

GRASP65
1.0 mg/mL
Western Blot 1:1000 - 1:5000, Immunoprecipitation 2 - 10 ug/mg
NP_114105.1
Golgi peripheral membrane protein p65, Golgi phosphoprotein 5MGC118897, golgi reassembly stacking protein 1, 65kDa, Golgi reassembly-stacking protein 1, Golgi reassembly-stacking protein of 65 kDa, GRASP65MGC118894, P6565 kDa
Rabbit
IgG
100 ul
Golgi Apparatus Markers
Primary
64689
Human, Mouse
Western Blot, Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
GORASP1
The immunogen recognized by this antibody maps to a region between residue 390 to 440 of human Golgi Reassembly Stacking Protein 1, 65kDa using the numbering given in entry NP_114105.1.
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
The epitope recognized by mapping to a region between residue 390 to 440 of human Golgi Reassembly Stacking Protein 1, 65kDa using the numbering given in entry NP_114105.1.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit