Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRIA1 (phospho S849), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic phosphopeptide of GRIA1.

Supplier:  Abnova Corporation PAB25848

Catalog No. 89-127-477


Add to Cart

Description

Description

Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. These receptors are heteromeric protein complexes with multiple subunits, each possessing transmembrane regions, and all arranged to form a ligand-gated ion channel. The classification of glutamate receptors is based on their activation by different pharmacologic agonists. This gene belongs to a family of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate (AMPA) receptors. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: Q-Q-Sp-I-N
Specifications

Specifications

GRIA1
Polyclonal
Unconjugated
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
GRIA1
GLUH1/GLUR1/GLURA/HBGR1/MGC133252
Rabbit
Affinity chromatography
RUO
2890
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Western Blot
1 mg/mL
Rabbit polyclonal antibody raised against synthetic phosphopeptide of GRIA1.
In PBS (without mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
P42261
GRIA1
Synthetic phosphopeptide corresponding to residues surrounding S849 of human GRIA1.
100 μg
Primary
Human, Mouse, Rat
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only