Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRIN2B Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of GRIN2B.

Manufacturer:  Abnova Corporation PAB12211

Catalog No. 89-115-672


Add to cart

Description

Description

N-methyl-D-aspartate (NMDA) receptors are a class of ionotropic glutamate receptors. NMDA receptor channel has been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of three different subunits: NR1 (GRIN1), NR2 (GRIN2A, GRIN2B, GRIN2C, or GRIN2D) and NR3 (GRIN3A or GRIN3B). The NR2 subunit acts as the agonist binding site for glutamate. This receptor is the predominant excitatory neurotransmitter receptor in the mammalian brain. [provided by RefSeq

Sequence: HVYEK
Specifications

Specifications

GRIN2B
1 mg/mL
Rabbit polyclonal antibody raised against synthetic peptide of GRIN2B.
In PBS (without mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
Q13224
MGC142178|MGC142180|NMDAR2B|NR2B|hNR3
Rabbit
Immunoaffinity purification
RUO
Primary
2904
Human, Murine, Rat
Immunofluorescence
Unconjugated
Immunofluorescence (1:100-1:200) The optimal working dilution should be determined by the end user.
GRIN2B
Liquid
GRIN2B
A synthetic peptide (conjugated with KLH) corresponding to residues surrounding Y1474 of human GRIN2B.
100 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
NMDAR2B (Ab-1472) antibody detects endogenous levels of total NMDAR2B protein.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only