Learn More
GRIN2B (phospho Y1474), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of GRIN2B.
Supplier: Abnova Corporation PAB12171
Description
N-methyl-D-aspartate (NMDA) receptors are a class of ionotropic glutamate receptors. NMDA receptor channel has been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of three different subunits: NR1 (GRIN1), NR2 (GRIN2A, GRIN2B, GRIN2C, or GRIN2D) and NR3 (GRIN3A or GRIN3B). The NR2 subunit acts as the agonist binding site for glutamate. This receptor is the predominant excitatory neurotransmitter receptor in the mammalian brain. [provided by RefSeq
Sequence: HVYpEKSpecifications
GRIN2B | |
Polyclonal | |
Unconjugated | |
Immunofluorescence (1:100-1:200) The optimal working dilution should be determined by the end user. | |
GRIN2B | |
MGC142178/MGC142180/NMDAR2B/NR2B/hNR3 | |
Rabbit | |
Immunoaffinity purification | |
RUO | |
2904 | |
Human, Mouse, Rat | |
Liquid |
Immunofluorescence | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of GRIN2B. | |
In PBS (without mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
Q13224 | |
GRIN2B | |
Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y1474 of human GRIN2B. | |
100 μL | |
Primary | |
NMDAR2B (phospho-Tyr1474) antibody detects endogenous levels of NMDAR2B only when phosphorylated at Tyr1474. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only