Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-GSAP, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP1784000.025ML

Catalog No. NBP17840025


Add to cart

Description

Description

GSAP Polyclonal antibody specifically detects GSAP in Human samples. It is validated for Western Blot, Simple Western.
Specifications

Specifications

GSAP
1.0 mg/mL
Western Blot 1:1000, Simple Western 1:100
A4D1B5
Gamma-secretase-activating protein, pigeon homolog (Drosophila)
Rabbit
IgG
0.025 ml
Alzheimers Research, Neurodegeneration, Neuroscience
Primary
54103
Western Blot
Unconjugated
PBS and 30% Glycerol with 0.05% Sodium Azide
Affinity Purified
PION
A synthetic peptide made to an internal portion of the human GSAP protein (between residues 150-200) [UniProt A4D1B5]
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit