Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-GSH2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243705

Catalog No. NBP243705


Add to cart

Description

Description

Specifically detects GSH2 in Human samples, and it is validated for Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blotting

GSH2 Polyclonal antibody specifically detects GSH2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

GSH2
Unconjugated
PBS (pH 7.0) and 20% Glycerol with 0.025% Proclin 300
GS homeobox 2, GSH2, Homeobox protein GSH-2
Rabbit
IgG
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500
Ion Exchange Chromatography
GSX2
Recombinant protein encompassing a sequence within the N-terminus region of human GSX2. The exact sequence is proprietary.
Immunogen affinity purified
RUO
Primary
170825
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only