Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSR Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against partial recombinant GSR.

Manufacturer:  Abnova Corporation PAB1207

Catalog No. 89-115-574


Add to cart

Description

Description

Specifications

Specifications

GSR
Unconjugated
Western Blot (1:1800) The optimal working dilution should be determined by the end user.
GSR
MGC78522
Rabbit
100 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Immunohistochemistry, Western Blot
Rabbit polyclonal antibody raised against partial recombinant GSR.
In HEPES, 150mM NaCl (50% glycerol, 0.01% BSA, 0.03% sodium azide)
Liquid
GSR
Recombinant protein corresponding to amino acids 44-522 of human GSR.
RUO
Primary
2936
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only