Learn More
GST Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against GST.
Manufacturer:  Abnova Corporation PAB13748
Catalog No. 89-137-671
Add to cart
Description
Description
Specifications
Specifications
GST
Unconjugated
ELISA (Detection: 1:5000-1:20000 Capture: 1:200-1:500) Western Blot (1:5000-1:10000) The optimal working dilution should be determined by the end user.
Liquid
Native purified GST.
RUO
Primary
Not Provided
ELISA, Immunoblot, Immunoprecipitation, Western Blot
Rabbit polyclonal antibody raised against GST.
In PBS, pH 7.2 (0.09% sodium azide)
Rabbit
1 mg
Store at 4°C.
Polyclonal