Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAVCR2 Rabbit anti-Chimpanzee, Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of HAVCR2.

Manufacturer:  Abnova Corporation PAB14656

Catalog No. 89-117-219


Add to cart

Description

Description

CD4 (MIM 186940)-positive T helper lymphocytes can be divided into types 1 (Th1) and 2 (Th2) on the basis of their cytokine secretion patterns. Th1 cells and their associated cytokines are involved in cell-mediated immunity to intracellular pathogens and delayed-type hypersensitivity reactions, whereas Th2 cells are involved in the control of extracellular helminthic infections and the promotion of atopic and allergic diseases. The 2 types of cells also cross-regulate the functions of the other. TIM3 is a Th1-specific cell surface protein that regulates macrophage activation and enhances the severity of experimental autoimmune encephalomyelitis in mice.[supplied by OMIM

Sequence: ISTLANELRDSRLANDLRD
Specifications

Specifications

HAVCR2
Unconjugated
ELISA (1:100-1:1000) The optimal working dilution should be determined by the end user.
HAVCR2
FLJ14428|KIM-3|TIM3|TIMD3|Tim-3
Rabbit
100 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
The amino acid sequence used as immunogen is 100% homologous in human and 94% in chimp.
ELISA
Rabbit polyclonal antibody raised against synthetic peptide of HAVCR2.
In serum (50% glycerol, 0.025% sodium azide)
Liquid
HAVCR2
A synthetic peptide corresponding to amino acids 176-194 of human HAVCR2.
RUO
Primary
84868
Chimpanzee, Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only