Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDLBP Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of HDLBP.

Manufacturer:  Abnova Corporation PAB14561

Catalog No. 89-117-143


Add to cart

Description

Description

High density lipoprotein-binding protein, also known as vigilin, is a 110kDa protein that specifically binds HDL molecules and may function in the removal of excess cellular cholesterol.[supplied by OMIM

Specifications

Specifications

HDLBP
Unconjugated
Western Blot (2 ug/ mL) The optimal working dilution should be determined by the end user.
HDLBP
FLJ16432|HBP|PRO2900|VGL
Rabbit
100 uL
Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of HDLBP.
In Tris-glycine, 150mM NaCl (0.05% sodium azide)
Liquid
HDLBP
A synthetic peptide corresponding to amino acids 100-180 of human HDLBP.
RUO
Primary
3069
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only