Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HECW1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant HECW1.

Manufacturer:  Abnova Corporation PAB20386

Catalog No. 89-121-887


Add to cart

Description

Description

Sequence: STEPESAQIQDSPMNNLMESGSGEPRSEAPESSESWKPEQLGEGSVPDGPGNQSIELSRPAEEAAVITEAGDQGMVSVGPEGAGELLAQVQKDIQPAPSAEELAEQLDLGEEASALLLEDGEAPAS
Specifications

Specifications

HECW1
Unconjugated
Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user.
HECW1
KIAA0322|NEDL1
Rabbit
IgG
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Immunohistochemistry, Immunohistochemistry (Formalin/Paraffin)
Rabbit polyclonal antibody raised against recombinant HECW1.
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Liquid
HECW1
Recombinant protein corresponding to amino acids of human HECW1.
Antigen affinity purification
RUO
Primary
23072
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only