Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Helicobacter pylori, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP1359570.1ML

Catalog No. NBP13595701


Add to cart

Description

Description

Helicobacter Pylori Polyclonal antibody specifically detects Helicobacter Pylori in Bacteria samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Helicobacter Pylori
Unconjugated
10mM PBS (pH 7.4) with Sodium Azide
H. pylori
Purified Helicobacter pylori
Immunogen affinity purified
RUO
Primary
This product is specific for H. Pylori
Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100
Affinity Purified
Rabbit
IgG
0.1 ml
Store at 4C. Do not freeze.
Polyclonal
Bacteria
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit