Learn More
hepatitis C virus core Polyprotein Rabbit anti-Viruses, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of hepatitis C virus core Polyprotein.
Manufacturer:  Abnova Corporation PAB16135
Catalog No. 89-138-195
Add to cart
Description
Description
Specifications
Specifications
hepatitis C virus core Polyprotein
Unconjugated
ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
A synthetic peptide corresponding to N-terminus of HCV core Polyprotein.
RUO
Primary
Virus
ELISA, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of hepatitis C virus core Polyprotein.
In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
Rabbit
100 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal