Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-HISPPD2A, (1383 to 1433), Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP244293

Catalog No. NBP244293


Add to cart

Description

Description

Specifically detects HISPPD2A in Human samples, and it is validated for Immunoprecipitation, Western Blotting

HISPPD2A Polyclonal antibody specifically detects HISPPD2A in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

HISPPD2A
1.0 mg/mL
Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg lysate
Affinity Purified
PPIP5K1
The epitope recognized by this antibody maps to a region between residue 1383 to 1433 of human Inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 1 using the numbering given in entry Q6PFW1.1 (GeneID 9677).
Immunogen affinity purified
RUO
Primary
9677
Western Blot, Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
diphosphoinositol pentakisphosphate kinase 1, histidine acid phosphatase domain-containing protein 2A, inositol pyrophosphate synthase 1, VIP1
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only