Learn More
HIST1H2BH Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of HIST1H2BH.
Manufacturer: Abnova Corporation PAB17428
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H2B family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6. [provided by RefSeq
Specifications
HIST1H2BH | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of HIST1H2BH. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
H2B/j/H2BFJ | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total HIST1H2BH protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) Immunofluorescence (1:500-1:1000) ELISA (1:5000) The optimal working dilution should be determined by the end user. | |
HIST1H2BH | |
HIST1H2BH | |
A synthetic peptide corresponding to internal of human HIST1H2BH. | |
RUO | |
8345 | |
Human, Mouse | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only