Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Rabbit anti-Histidine Tag, Clone: RM146,
Rabbit Monoclonal Antibody
Supplier: MilliporeSigma™ 041664
Description
Specifically detects Histidine Tag clone: RM146 in All samples, and it is validated for Immunoprecipitation, Western Blotting
Specifications
Histidine Tag | |
Monoclonal | |
Rabbit monoclonal in PBS with 1% BSA and 0.09% sodium azide. | |
Mixture of a peptide with 6xHis-Tag at the N-terminus and another peptide with 6xHis-Tag at the C-terminus. | |
100 μg | |
Secondary and Control Antibodies | |
All species | |
Purified |
Immunoprecipitation, Western Blot | |
RM146 | |
Rabbit | |
Protein A Purfied | |
RUO | |
Secondary | |
Stable for one year at 2°C to 8°C from date of receipt. Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only