Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
histone H2B, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of histone H2B.
Manufacturer: Abnova Corporation PAB16672
Description
Sequence: SAAEKKPASKAPAGKASpecifications
histone H2B | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of histone H2B. | |
In antiserum (0.05% sodium azide) | |
htb1 | |
A synthetic peptide corresponding to Schizosaccharomyces pombe histone H2B. | |
RUO | |
2539523 | |
Liquid |
ChIP assay, Western Blot | |
Unconjugated | |
Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
htb1 | |
Rabbit | |
50 μL | |
Primary | |
S. pombe |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only