Learn More
Histone H3 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Histone H3.
Manufacturer:  Abnova Corporation PAB17295
Catalog No. 89-138-331
Add to cart
Description
Description
Specifications
Specifications
Histone H3
Unconjugated
Immunohistochemistry (1:50-1:100) ELISA (1:5000) The optimal working dilution should be determined by the end user.
Liquid
A synthetic peptide corresponding to internal of human Histone H3.
RUO
Primary
This antibody detects endogenous levels of total Histone H3 protein.
Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against synthetic peptide of Histone H3.
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
Rabbit
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine, Rat