Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV-1 Gag p15 protein, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant HIV-1 Gag p15 protein.

Manufacturer:  Abnova Corporation PAB1180

Catalog No. 89-137-478

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

HIV-1 Gag p15 protein
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Recombinant protein corresponding to human immunodeficiency virus type 1 Gag p15 protein.
RUO
Primary
ELISA, Western Blot
Rabbit polyclonal antibody raised against recombinant HIV-1 Gag p15 protein.
In buffer containing 0.09% sodium azide
Rabbit
50 uL
Store at 4°C.
Polyclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only