Learn More
HK2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of HK2.
Supplier:  Abnova Corporation PAB4004
Catalog No. 89-131-229
Add to Cart
Description
Description
Hexokinases phosphorylate glucose to produce glucose-6-phosphate, the first step in most glucose metabolism pathways. This gene encodes hexokinase 2, the predominant form found in skeletal muscle. It localizes to the outer membrane of mitochondria. Expression of this gene is insulin-responsive, and studies in rat suggest that it is involved in the increased rate of glycolysis seen in rapidly growing cancer cells. [provided by RefSeq
Specifications
Specifications
HK2
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of HK2.
In PBS (0.09% sodium azide)
DKFZp686M1669/HKII/HXK2
Rabbit
Protein G purification
RUO
3099
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user.
HK2
HK2
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human HK2.
400 μL
Primary
Human
Liquid
Product Suggestions
Product Suggestions