Learn More
HMGN2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of HMGN2.
Manufacturer:  Abnova Corporation PAB18390
Catalog No. 89-119-954
Add to cart
Description
Description
Specifications
Specifications
HMGN2
1 mg/mL
Rabbit polyclonal antibody raised against synthetic peptide of HMGN2.
In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
Liquid
HMGN2
A synthetic peptide corresponding to human HMGN2.
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
This antibody is specific to HMGN2.
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (1:50-1:100) Immunofluorescence (1:500-1:1000) ELISA (1:5000) The optimal working dilution should be determined by the end user.
HMGN2
HMG17|MGC5629|MGC88718
Rabbit
Affinity purification
RUO
Primary
3151
Human, Murine, Rat