Learn More
HNRPLL Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of HNRPLL.
Manufacturer: Abnova Corporation PAB17745
Description
HNRNPLL is a master regulator of activation-induced alternative splicing in T cells. In particular, it alters splicing of CD45 (PTPRC; MIM 151460), a tyrosine phosphatase essential for T-cell development and activation (Oberdoerffer et al., 2008 [PubMed 18669861]).[supplied by OMIM
Specifications
HNRPLL | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of HNRPLL. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
SRRF | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total HNRPLL protein. | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) ELISA (1:20000) The optimal working dilution should be determined by the end user. | |
HNRPLL | |
HNRPLL | |
A synthetic peptide corresponding to internal of human HNRPLL. | |
RUO | |
92906 | |
Human, Murine |
Safety and Handling
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only