Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-HOOK3, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP244279

Catalog No. NBP244279


Add to cart

Description

Description

Specifically detects HOOK3 in Human, Mouse samples, and it is validated for Immunoprecipitation, Western Blotting

HOOK3 Polyclonal antibody specifically detects HOOK3 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

HOOK3
1.0 mg/mL
Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg lysate
Affinity Purified
HOOK3
The epitope recognized by this antibody maps to a region between residue 425 to 475 of human Protein Hook homolog 3 using the numbering given in entry Q86VS8.2 (GeneID 84376).
Immunogen affinity purified
RUO
Primary
84376
Western Blot, Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
FLJ31058, hHK3, HK3h-hook3, hook homolog 3 (Drosophila), protein Hook homolog 3
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only