Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HTR1D Rabbit anti-Human, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic peptide of HTR1D.
Manufacturer: Abnova Corporation PAB25571
Specifications
HTR1D | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (17 ug/mL) The optimal working dilution should be determined by the end user. | |
HTR1D | |
5-HT1D|HT1DA|HTR1DA|HTRL|RDC4 | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
Primary | |
3352 | |
Human |
Immunohistochemistry (PFA fixed) | |
Rabbit polyclonal antibody raised against synthetic peptide of HTR1D. | |
In PBS (0.09% sodium azide) | |
Liquid | |
HTR1D | |
A synthetic peptide corresponding to 16 amino acids at N-terminal extracellular domain of human HTR1D. | |
50 ug | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only