Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HTR1E Rabbit anti-Guinea pig, Horse, Human, Monkey, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of HTR1E.
Supplier: Abnova Corporation PAB16399
Specifications
HTR1E | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of HTR1E. | |
In PBS (0.09% sodium azide) | |
5-HT1E | |
Rabbit | |
Immunoaffinity purification | |
RUO | |
3354 | |
Equine, Guinea Pig, Human, Monkey | |
Liquid |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (16-24 ug/mL) The optimal working dilution should be determined by the end user. | |
HTR1E | |
HTR1E | |
A synthetic peptide (conjugated with KLH) corresponding to human HTR1E. | |
50 μg | |
Primary | |
3rd Cytoplasmic domain of human. | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction