Learn More
IgA Rabbit anti-Human, FITC, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Manufacturer:  Invitrogen SA136096
Catalog No. 01-671-508
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This antibody shows no cross reactivity with albumin fractions of human serum.
Thermo Scientific Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
Human IgA
Polyclonal
FITC
Liquid
Immunoglobulins.
Ion-Exchange Chromatography, Size-Exclusion - Gel Filtration
RUO
Secondary Antibody
Polyclonal
Immunohistochemistry
1.2 mg/mL
PBS with 50% glycerol and no preservative; pH 7.5
Rabbit
IgG
1 mL
4° C, store in dark
Secondary
Human