Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
IgG (H+L) Rabbit anti-Human, FITC, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Manufacturer:  Invitrogen SA136099
Catalog No. 01-671-511
Description
Description
This antibody shows no cross reactivity with albumin fractions of human serum.
Thermo Scientific Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
Human IgG (H+L)
Polyclonal
FITC
Liquid
Immunoglobulins.
Purified
RUO
Secondary Antibody
Polyclonal
Immunohistochemistry
1.2 mg/mL
PBS with 50% glycerol and no preservative; pH 7.5
Rabbit
IgG
1 mL
4° C, store in dark
Secondary
Human