Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Huntingtin (p Ser1201), Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP244146

Catalog No. NBP244146


Add to cart

Description

Description

Specifically detects Huntingtin (p Ser1201) in Human, Mouse, Rat samples, and it is validated for ELISA, Western Blotting

Huntingtin p Ser1201 Polyclonal antibody specifically detects Huntingtin p Ser1201 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Huntingtin (p Ser1201)
1.08 mg/mL
Western Blot 1ug/ml, ELISA 1: 10000
Affinity Purified
HTT
Anti-Huntington [p Ser1201] affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to the internal region of human Huntingtin protein.
Immunogen affinity purified
RUO
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
Western Blot, ELISA
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 50% (v/v) Glycerol with 1 mg/ml Bovine Serum Albumin (BSA) with No Preservative
HD protein, huntingtin, huntingtin (Huntington disease), IT15HDHuntington disease protein
Rabbit
IgG
0.1 mg
Apoptosis, Autophagy, Cytoskeleton Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific
Primary
3064
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only