Learn More
Helicobacter pylori Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against Helicobacter pylori.
Manufacturer:  Abnova Corporation PAB11934
Catalog No. 89-137-482
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
Helicobacter pylori
Lot specific
Rabbit polyclonal antibody raised against Helicobacter pylori.
In 10mM PBS, pH 7.4 (1% BSA, 0.09% sodium azide)
Rabbit
Immunoaffinity purification
RUO
Primary
Helicobacter pylori
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user.
Liquid
Purified Helicobacter pylori.
100 uL
Store at 4°C.
Polyclonal