Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Icam1, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide surrounding amino acid 530 of Icam1.

Manufacturer:  Abnova Corporation PAB14893

Catalog No. 89-117-337


Add to cart

Description

Description

Specifications

Specifications

Icam1
Unconjugated
Western Blot (0.5-4 ug/mL) The optimal working dilution should be determined by the end user.
Icam1
CD54|ICAM
Rabbit
100 ug
Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
This antibody recognizes ~89-92 KDa ICAM-1.
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide surrounding amino acid 530 of Icam1.
In PBS (30% glycerol 0.5% BSA, 0.01% thimerosal)
Liquid
Icam1
A synthetic peptide corresponding to residues surrounding 530 of rat Icam1.
RUO
Primary
25464
Human, Murine, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only