Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgA Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of IgA.

Manufacturer:  Abnova Corporation PAB25169

Catalog No. 89-138-409


Add to cart

Description

Description

Specifications

Specifications

IgA
1 mg/mL
Rabbit polyclonal antibody raised against synthetic peptide of IgA.
In PBS, pH 7.2 (0.05% sodium azide)
Rabbit
Affinity purification
RUO
Primary
IgA polyclonal antibody detects endogenous levels of IgA protein.
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
Liquid
A synthetic peptide corresponding to IgA.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only