Learn More
IgA2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native IgA2.
Manufacturer:  Abnova Corporation PAB19859
Catalog No. 89-138-384
Add to cart
Description
Description
Specifications
Specifications
IgA2
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Native purified IgA2 from human plasma.
150 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
ELISA, Western Blot
Rabbit polyclonal antibody raised against native IgA2.
In PBS, pH 7.5 (0.02% sodium azide, 50% glycerol (v/v))
Rabbit
Affinity purification
RUO
Primary
Human