Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF1R Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against partial recombinant IGF1R.

Manufacturer:  Abnova Corporation PAB16190

Catalog No. 89-118-080


Add to cart

Description

Description

This receptor binds insulin-like growth factor with a high affinity. It has tyrosine kinase activity. The insulin-like growth factor I receptor plays a critical role in transformation events. Cleavage of the precursor generates alpha and beta subunits. It is highly overexpressed in most malignant tissues where it functions as an anti-apoptotic agent by enhancing cell survival. [provided by RefSeq

Specifications

Specifications

IGF1R
Unconjugated
ELISA (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
IGF1R
CD221|IGFIR|JTK13|MGC142170|MGC142172|MGC18216
Rabbit
100 ug
Store at -20°C on dry atmosphere. After reconstitution with deionized water
Polyclonal
Human
ELISA
Rabbit polyclonal antibody raised against partial recombinant IGF1R.
Lyophilized from PBS.
Lyophilized
IGF1R
Recombinant protein corresponding to extracellular region of human IGF1R.
RUO
Primary
3480
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only